Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
neto2
Entrez Gene ID
780374
Gene Name
neuropilin and tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.4: 25278317 - 25302668 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079444.1 NP_001072912.1 3 - 4 567 86
GenBank mRNA BC121922.1 - 3 - 4 567 86

View Full Product Details