The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079451.1 | NP_001072919.1 | 7 - 8 | 553 | 62 |
XM_012970593.1 | XP_012826047.1 | 7 - 8 | 787 | 62 | |
GenBank mRNA | BC121427.1 | - | 7 - 8 | 553 | 62 |