Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kremen2
Entrez Gene ID
780393
Gene Name
kringle containing transmembrane protein 2
Gene Aliases
krm2
Chromosome Location
Chr.9: 7094456 - 7105302 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079463.1 NP_001072931.1 3 - 4 524 101
GenBank mRNA BC124059.1 - 3 - 4 524 101

View Full Product Details