Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tma16
Entrez Gene ID
549385
Gene Name
translation machinery associated 16 homolog
Gene Aliases
c4orf43
Chromosome Location
Chr.1: 42388191 - 42423317 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016631.3 NP_001016631.1 6 - 7 531 115
XM_012962584.2 XP_012818038.1 7 - 8 846 115
GenBank mRNA BC135799.1 - 6 - 7 531 115
CR760447.2 - 6 - 7 438 115

View Full Product Details