Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vsig10
Entrez Gene ID
733840
Gene Name
V-set and immunoglobulin domain containing 10
Gene Aliases
-
Chromosome Location
Chr.1: 156722746 - 156737110 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045591.1 NP_001039056.1 4 - 5 1144 82
XM_012964268.2 XP_012819722.1 4 - 5 1432 82
GenBank mRNA CR848573.2 - 4 - 5 1144 82

View Full Product Details