Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vsig10
Entrez Gene ID
733840
Gene Name
V-set and immunoglobulin domain containing 10
Gene Aliases
-
Chromosome Location
Chr.1: 156722746 - 156737110 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045591.1 NP_001039056.1 8 - 9 1746 80
GenBank mRNA CR848573.2 - 8 - 9 1746 80

View Full Product Details