Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kcnn1
Entrez Gene ID
779939
Gene Name
potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1
Gene Aliases
hsk1, sk1, skca1
Chromosome Location
Chr.1: 99395054 - 99469441 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079016.1 NP_001072484.1 2 - 3 540 87
XM_012965355.2 XP_012820809.1 2 - 3 528 87
XM_012965371.2 XP_012820825.1 2 - 3 528 87
GenBank mRNA BC121911.1 - 2 - 3 540 87

View Full Product Details