Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stmn4
Entrez Gene ID
779944
Gene Name
stathmin 4
Gene Aliases
-
Chromosome Location
Chr.5: 146422785 - 146427746 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079021.1 NP_001072489.1 - 586 80
GenBank mRNA BC121918.1 - - 586 80

View Full Product Details