Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
spon2
Entrez Gene ID
780015
Gene Name
spondin 2, extracellular matrix protein
Gene Aliases
-
Chromosome Location
Chr.1: 14487126 - 14498894 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079092.2 NP_001072560.2 - 833 74
GenBank mRNA DR870839.1 - 3 - 4 464 74

View Full Product Details