Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbr1
Entrez Gene ID
780042
Gene Name
T-box, brain 1
Gene Aliases
T-brain-1, tbr-1, tes-56, xtbr1
Chromosome Location
Chr.9: 67216122 - 67230387 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079119.1 NP_001072587.1 3 - 4 1132 102
XM_012970558.2 XP_012826012.1 3 - 4 1165 102
GenBank mRNA BC122952.1 - 3 - 4 1132 102
BC136086.1 - 3 - 4 1004 102

View Full Product Details