Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c11orf49
Entrez Gene ID
780244
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
Chromosome Location
Chr.4: 151404 - 241262 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079316.1 NP_001072784.1 5 - 6 473 73
XM_012954043.2 XP_012809497.1 4 - 5 661 73
GenBank mRNA BC118860.1 - 5 - 6 473 73

View Full Product Details