Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
spata5l1
Entrez Gene ID
780248
Gene Name
spermatogenesis associated 5-like 1
Gene Aliases
-
Chromosome Location
Chr.3: 94597460 - 94637896 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079319.1 NP_001072787.1 4 - 5 1507 95
XM_012959288.2 XP_012814742.1 4 - 5 1559 95
XM_012959289.2 XP_012814743.1 4 - 5 1523 95
XM_012959290.2 XP_012814744.1 6 - 7 1638 95
XM_012959291.2 XP_012814745.1 4 - 5 1559 95
XM_012959292.2 XP_012814746.1 4 - 5 1559 95
XM_018092183.1 XP_017947672.1 4 - 5 1559 95
GenBank mRNA BC121246.1 - 4 - 5 1507 95

View Full Product Details