Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hpcal4
Entrez Gene ID
780259
Gene Name
hippocalcin like 4
Gene Aliases
hlp4
Chromosome Location
Chr.2: 86861073 - 86878090 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079330.1 NP_001072798.1 2 - 3 240 77
GenBank mRNA BC121367.1 - 2 - 3 240 77

View Full Product Details