Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cracr2b
Entrez Gene ID
100036622
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
efcab4a
Chromosome Location
Chr.4: 35116399 - 35131049 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097181.1 NP_001090650.1 5 - 6 5741 92
GenBank mRNA BC127278.1 - 5 - 6 5741 92
CR761002.2 - 8 - 9 1060 92

View Full Product Details