Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tet3
Entrez Gene ID
100036628
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
xlTet3
Chromosome Location
Chr.3: 112974754 - 113023824 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097187.1 NP_001090656.1 6 - 7 3715 86
GenBank mRNA BC127289.1 - 6 - 7 3715 86

View Full Product Details