Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bid
Entrez Gene ID
100036674
Gene Name
BH3 interacting domain death agonist
Gene Aliases
fp497, xbid
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097226.1 NP_001090695.1 - 355 59
GenBank mRNA EF065099.1 - - 355 59

View Full Product Details