Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
p3h1
Entrez Gene ID
100036679
Gene Name
prolyl 3-hydroxylase 1
Gene Aliases
lepre1, leprecan
Chromosome Location
Chr.7: 68525212 - 68553379 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097231.1 NP_001090700.1 11 - 12 1756 79
XM_018095197.1 XP_017950686.1 11 - 12 2058 79
GenBank mRNA BC128622.1 - 11 - 12 1756 79

Gene Symbol
LOC101734450
Entrez Gene ID
101734450
Gene Name
prolyl 3-hydroxylase 1-like
Gene Aliases
-
Chromosome Location
Chr.7: 68431861 - 68436591 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018095675.1 XP_017951164.1 2 - 3 314 79

Gene Symbol
LOC101734514
Entrez Gene ID
101734514
Gene Name
prolyl 3-hydroxylase 1-like
Gene Aliases
-
Chromosome Location
Chr.7: 68448964 - 68476140 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018095677.1 XP_017951166.1 8 - 9 785 79

Gene Symbol
LOC101734646
Entrez Gene ID
101734646
Gene Name
prolyl 3-hydroxylase 1-like
Gene Aliases
-
Chromosome Location
Chr.7: 68499547 - 68521704 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012967249.2 XP_012822703.1 7 - 8 818 79

Gene Symbol
LOC105945914
Entrez Gene ID
105945914
Gene Name
prolyl 3-hydroxylase 1-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016684009: 20138 - 23798 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012954954.2 XP_012810408.1 2 - 3 353 79

View Full Product Details