Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sept2
Entrez Gene ID
100036706
Gene Name
septin 2
Gene Aliases
Septin-2B, diff6, hnedd5, nedd5, pnutl3, sept2-B, sept2-a
Chromosome Location
Chr.5: 122320253 - 122338728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097254.1 NP_001090723.1 8 - 9 872 86
XM_012969927.2 XP_012825381.1 9 - 10 999 86
XM_012969928.2 XP_012825382.1 9 - 10 956 86
GenBank mRNA BC127290.1 - 8 - 9 872 86
CT030346.1 - 8 - 9 870 86

View Full Product Details