Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gip
Entrez Gene ID
100036740
Gene Name
gastric inhibitory polypeptide
Gene Aliases
xgip
Chromosome Location
Chr.10: 5270214 - 5277755 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097282.1 NP_001090751.1 3 - 4 263 69
XM_012971487.1 XP_012826941.1 3 - 4 304 69
GenBank mRNA EF010532.1 - 3 - 4 263 69

View Full Product Details