Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ogfrl1
Entrez Gene ID
100037841
Gene Name
opioid growth factor receptor-like 1
Gene Aliases
-
Chromosome Location
Chr.5: 91584603 - 91616693 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097287.1 NP_001090756.1 3 - 4 460 113
XM_012962530.2 XP_012817984.1 3 - 4 760 113
XM_018093724.1 XP_017949213.1 3 - 4 261 113
GenBank mRNA BC128651.1 - 3 - 4 460 113

View Full Product Details