Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syt8
Entrez Gene ID
100037878
Gene Name
synaptotagmin 8
Gene Aliases
-
Chromosome Location
Chr.4: 12526841 - 12543725 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097318.1 NP_001090787.1 4 - 5 415 78
XM_012960380.2 XP_012815834.1 4 - 5 481 78
XM_012960381.2 XP_012815835.1 3 - 4 432 78
GenBank mRNA BC135170.1 - 4 - 5 415 78

View Full Product Details