Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
paip1
Entrez Gene ID
100038266
Gene Name
poly(A) binding protein interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 181953332 - 181972153 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097384.1 NP_001090853.1 3 - 4 676 70
GenBank mRNA BC135141.1 - 3 - 4 676 70

View Full Product Details