Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab31
Entrez Gene ID
100101725
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
rab22b
Chromosome Location
Chr.6: 89517800 - 89537123 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100241.1 NP_001093711.1 4 - 5 343 101
GenBank mRNA BC135941.1 - 4 - 5 343 101

View Full Product Details