Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rac3
Entrez Gene ID
100101726
Gene Name
Rac family small GTPase 3
Gene Aliases
-
Chromosome Location
Chr.10: 23533515 - 23546674 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100242.1 NP_001093712.1 3 - 4 304 62
GenBank mRNA BC135751.1 - 3 - 4 304 62

View Full Product Details