Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sh2b3
Entrez Gene ID
100101733
Gene Name
SH2B adaptor protein 3
Gene Aliases
celiac13, lnk
Chromosome Location
Chr.1: 154509714 - 154604541 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100247.1 NP_001093717.1 7 - 8 1851 103
GenBank mRNA BC134803.1 - 7 - 8 1851 103

View Full Product Details