Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fscn2
Entrez Gene ID
100101745
Gene Name
fascin actin-bundling protein 2, retinal
Gene Aliases
rfsn, rp30
Chromosome Location
Chr.10: 20601829 - 20627582 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100254.1 NP_001093724.1 4 - 5 1341 134
GenBank mRNA BC135752.1 - 4 - 5 1341 134

View Full Product Details