Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kazald1
Entrez Gene ID
100101767
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
Xigfbp-rp10, ba108l7.1, bono1, fksg28, fksg40, igfbp-rp10
Chromosome Location
Chr.7: 21729956 - 21760844 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100263.1 NP_001093733.1 2 - 3 745 102
GenBank mRNA BC135810.1 - 2 - 3 745 102

View Full Product Details