Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmeff1
Entrez Gene ID
100101772
Gene Name
transmembrane protein with EGF like and two follistatin like domains 1
Gene Aliases
X7365, h7365
Chromosome Location
Chr.6: 61673156 - 61814819 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100268.3 NP_001093738.2 2 - 3 796 85
XM_012964170.2 XP_012819624.1 2 - 3 846 85
GenBank mRNA BC135422.2 - 2 - 3 630 85

View Full Product Details