Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab3b
Entrez Gene ID
100101786
Gene Name
RAB3B, member RAS oncogene family
Gene Aliases
rab-3b
Chromosome Location
Chr.4: 86312180 - 86372573 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100276.1 NP_001093746.1 3 - 4 478 105
XM_018092876.1 XP_017948365.1 3 - 4 421 105
GenBank mRNA BC135348.1 - 3 - 4 478 105

View Full Product Details