Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rel
Entrez Gene ID
100124724
Gene Name
v-rel avian reticuloendotheliosis viral oncogene homolog
Gene Aliases
Xrel2, Xrel3, c-rel, rel2, rel3, v-rel, xrel
Chromosome Location
Chr.5: 27542611 - 27580390 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102707.1 NP_001096177.1 6 - 7 964 89
GenBank mRNA BC136176.1 - 6 - 7 964 89

View Full Product Details