Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
myl4
Entrez Gene ID
100124786
Gene Name
myosin light chain 4
Gene Aliases
MLC1av
Chromosome Location
Chr.10: 7174109 - 7181987 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102762.1 NP_001096232.1 3 - 4 342 86
GenBank mRNA BC135340.1 - 3 - 4 342 86
BC155857.1 - 3 - 4 320 86

View Full Product Details