Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
angptl7
Entrez Gene ID
100124808
Gene Name
angiopoietin like 7
Gene Aliases
-
Chromosome Location
Chr.7: 39862530 - 39866022 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102778.1 NP_001096248.1 2 - 3 604 72
GenBank mRNA BC135402.1 - 2 - 3 604 72

View Full Product Details