Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klhl30
Entrez Gene ID
100124817
Gene Name
kelch like family member 30
Gene Aliases
-
Chromosome Location
Chr.5: 122780213 - 122794058 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102785.1 NP_001096255.1 7 - 8 1596 102
XM_012969954.1 XP_012825408.1 7 - 8 1579 102
GenBank mRNA BC135429.1 - 7 - 8 1596 102

View Full Product Details