Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttc30a
Entrez Gene ID
100124881
Gene Name
tetratricopeptide repeat domain 30a
Gene Aliases
flr, ttc30
Chromosome Location
Chr.6: 71348470 - 71372426 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102833.1 NP_001096303.1 11 - 12 1189 72
GenBank mRNA BC135644.1 - 11 - 12 1189 72

View Full Product Details