Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cbx4
Entrez Gene ID
100124911
Gene Name
chromobox 4
Gene Aliases
XPolycomb, hpc2, nbp16, pc2
Chromosome Location
Chr.10: 24887757 - 24894064 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102857.1 NP_001096327.1 1 - 2 381 71
GenBank mRNA BC135729.1 - 1 - 2 381 71
CR760648.2 - 1 - 2 393 71
CU075495.1 - 1 - 2 372 71

View Full Product Details