Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nova1
Entrez Gene ID
100124925
Gene Name
neuro-oncological ventral antigen 1
Gene Aliases
-
Chromosome Location
Chr.8: 91767287 - 91822183 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102869.1 NP_001096339.1 1 - 2 345 70
XM_012967885.2 XP_012823339.1 1 - 2 780 70
GenBank mRNA BC135781.1 - 1 - 2 345 70

View Full Product Details