Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plppr1
Entrez Gene ID
100124964
Gene Name
phospholipid phosphatase related 1
Gene Aliases
lppr1
Chromosome Location
Chr.1: 104818063 - 104884809 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102900.1 NP_001096370.1 5 - 6 522 98
XM_012964809.2 XP_012820263.1 4 - 5 768 98
GenBank mRNA BC135902.1 - 5 - 6 522 98

View Full Product Details