Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem245
Entrez Gene ID
100124972
Gene Name
transmembrane protein 245
Gene Aliases
c9orf5
Chromosome Location
Chr.Un|NW_016683380: 1092628 - 1347775 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012953820.2 XP_012809274.1 1 - 2 2056 90
XM_018089411.1 XP_017944900.1 1 - 2 2055 90
GenBank mRNA BC135929.1 - 1 - 2 678 90

View Full Product Details