Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
maneal
Entrez Gene ID
100124973
Gene Name
mannosidase, endo-alpha-like
Gene Aliases
manea
Chromosome Location
Chr.2: 53756393 - 53782265 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102908.1 NP_001096378.1 4 - 5 825 82
XM_012956294.2 XP_012811748.1 4 - 5 1433 82
GenBank mRNA BC135932.1 - 4 - 5 825 82

View Full Product Details