Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slitrk1
Entrez Gene ID
100124997
Gene Name
SLIT and NTRK like family member 1
Gene Aliases
-
Chromosome Location
Chr.2: 121164287 - 121167050 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102925.1 NP_001096395.1 1 - 1 2447 82
GenBank mRNA BC136007.1 - 1 - 1 2447 82

View Full Product Details