The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102934.1 | NP_001096404.1 | 2 - 3 | 971 | 80 |
XM_018092893.1 | XP_017948382.1 | 2 - 3 | 1366 | 80 | |
GenBank mRNA | BC136043.1 | - | 2 - 3 | 971 | 80 |