Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
peli2
Entrez Gene ID
100125008
Gene Name
pellino E3 ubiquitin protein ligase family member 2
Gene Aliases
pellino2, xpellino2
Chromosome Location
Chr.5: 28826203 - 28858485 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102936.1 NP_001096406.1 6 - 7 1038 80
XM_012962623.2 XP_012818077.1 6 - 7 2833 80
XM_012962624.2 XP_012818078.1 6 - 7 780 80
XM_012962625.2 XP_012818079.1 6 - 7 3004 80
XM_012962626.2 XP_012818080.1 6 - 7 1627 80
XM_018093761.1 XP_017949250.1 7 - 8 3097 80
GenBank mRNA BC136048.1 - 6 - 7 1038 80

View Full Product Details