Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zbtb49
Entrez Gene ID
100125011
Gene Name
zinc finger and BTB domain containing 49
Gene Aliases
znf509
Chromosome Location
Chr.1: 18593432 - 18607542 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102939.1 NP_001096409.1 2 - 3 204 167
GenBank mRNA BC136052.1 - 2 - 3 204 167

View Full Product Details