Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
brox
Entrez Gene ID
100125017
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
c1orf58
Chromosome Location
Chr.5: 5879538 - 5899846 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102942.1 NP_001096412.1 2 - 3 224 60
XM_018093762.1 XP_017949251.1 2 - 3 257 60
GenBank mRNA BC136067.1 - 2 - 3 224 60
CU075601.1 - 2 - 3 189 60

View Full Product Details