Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
brox
Entrez Gene ID
100125017
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
c1orf58
Chromosome Location
Chr.5: 5879538 - 5899846 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102942.1 NP_001096412.1 6 - 7 596 66
XM_018093762.1 XP_017949251.1 6 - 7 629 66
GenBank mRNA BC136067.1 - 6 - 7 596 66
CU075601.1 - 6 - 7 561 66

View Full Product Details