The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102944.1 | NP_001096414.1 | 2 - 3 | 1455 | 93 |
GenBank mRNA | BC136072.1 | - | 2 - 3 | 1455 | 93 |