Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
foxo3
Entrez Gene ID
100125025
Gene Name
forkhead box O3
Gene Aliases
foxo3a, xfoxo3
Chromosome Location
Chr.5: 75299516 - 75368665 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102949.1 NP_001096419.1 1 - 2 976 70
GenBank mRNA BC136083.1 - 1 - 2 976 70

View Full Product Details