Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a18
Entrez Gene ID
100125036
Gene Name
solute carrier family 25 member 18
Gene Aliases
gc2
Chromosome Location
Chr.Un|NW_016683387: 428047 - 449767 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102958.1 NP_001096428.1 3 - 4 392 70
XM_012958457.2 XP_012813911.1 4 - 5 469 70
XM_012958458.2 XP_012813912.1 4 - 5 459 70
XM_012958459.2 XP_012813913.1 3 - 4 416 70
GenBank mRNA BC136106.1 - 3 - 4 392 70

View Full Product Details