Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ehd4
Entrez Gene ID
100125054
Gene Name
EH domain containing 4
Gene Aliases
past4
Chromosome Location
Chr.8: 84478834 - 84510365 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102972.3 NP_001096442.2 4 - 5 1282 103
GenBank mRNA BC136141.1 - 4 - 5 1282 103
DN067449.1 - 1 - 2 325 103

View Full Product Details