Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tppp3
Entrez Gene ID
100125085
Gene Name
tubulin polymerization-promoting protein family member 3
Gene Aliases
-
Chromosome Location
Chr.4: 51871839 - 51887905 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102996.1 NP_001096466.1 2 - 3 440 96
XM_012960482.2 XP_012815936.1 3 - 4 559 96
XM_012960484.2 XP_012815938.1 2 - 3 482 96
GenBank mRNA BC136199.1 - 2 - 3 440 96

View Full Product Details